Principaux documents
CLS3779
Corning® microcarriers
enhanced attachment, weight 10 g
Synonyme(s) :
cell culture flasks, tissue culture flasks
10 G
139,00 €
139,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 G
139,00 €
About This Item
Code UNSPSC :
41122100
Nomenclature NACRES :
NB.13
139,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
enhanced attachment
Conditionnement
pack of 1
Fabricant/nom de marque
Corning 3779
Poids
10 g
Description générale
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
  • USP Class VI polystyrene material provides a consistent platform
  • Level (SAL) of 10-6
  • Certified nonpyrogenic
The enhanced attachment surface treatment infuses the surface of the microcarriers with oxygen, improving cell attachment, maximizing cell yield and viability. Also available as a closed system solution.
Informations légales
Corning is a registered trademark of Corning, Inc.