CLS3561
a
disposable spinner flask, polystyrene, non-treated, 1 L, w/ two sidearms, sterile, 6/cs
Synonyme(s) :
cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
1 480,00 €
About This Item
1 480,00 €
Produits recommandés
Matériaux
high-density polyethylene cap
polystyrene flask
screw top cap
Stérilité
sterile
Caractéristiques
cap
closure type Screw cap
two sidearms
Conditionnement
case of 6
Fabricant/nom de marque
Corning 3561
Capacité
1 L
Diam. du bras latéral
54 mm
Volume de travail
1-3 L
1000-3000 mL
Raccords
cap for GL 45
GL 45 neck
Adéquation
suitable for (scale-up cell culture; production of secreted proteins)
Catégories apparentées
Description générale
Convenience
The Corning disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
Confidence
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Compatibility
Made of ISO 10993 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Performance
The paddle size and height is optimized for each vessel size. A unique integrated magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface.
Conclusions
- Disposable large volume spinner flasks are equivalent to conventional glass spinner flasks for cell growth and viability
- A variety of cell types can be cultured in large volume disposable spinner flasks
- Disposable spinner flasks maintain equal aeration as compared to standard glass spinner flasks
- Cultured cells from glass and disposable spinner flasks are metabolically equivalent
- Culured cells in disposable spinner flasks product equal amounts of secreted proteins as cells cultured in traditional glass spinner flasks
Les clients ont également consulté
Articles
PLTMax® Human Platelet Lysate (hPL) is a superior serum-free and xeno-free media supplement alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) cultures.