Principaux documents
BAF189050011
Scienceware® autoclavable microsample tube rack
Lid Only, polypropylene
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.22
Matériaux
polypropylene
Caractéristiques
50 x 1.5ml tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18905-0011
Technique(s)
cryopreservation: suitable
L × l × H
25.4 cm (3.6 in.) × 13.3 cm (3 in.) × 7.3 cm (0.78 in.)
Description générale
Tray with rack insert can accommodate up to fifty 1.5ml tubes (or other tubes with a diameter less than 12mm) for large volume benchtop processing. To further protect tubes during transport or storage while also maintaining stackability, an optional polypropylene cover is available.
Caractéristiques et avantages
  • Deep tray holds more ice and water for extended use time
  • Includes flat perimeter rim for labeling tape
  • Can be safely stacked with identical racks when loaded or empty
  • Sturdy polypropylene plastic provides long life and heavy usage
  • Steam autoclavable at 121ºC (250ºF)
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP