Documents
AXYPDW20CS
Deep well plate
size 96 wells, size 2.0 mL, pkg of 10x5plates/cs, sterile
Synonyme(s) :
Deep well plate, Multi-well plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
clear bottom
conical bottom wells
Stérilité
sterile
Caractéristiques
lid: no
skirt
Fabricant/nom de marque
Axygen Inc P-DW-20-C-S
Corning
Conditionnement
pkg of 10x5plates/cs
Taille
2.0 mL
96 wells
Description générale
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
Informations légales
Axygen is a registered trademark of Corning, Inc.