Principaux documents
Z801321
Hellma® absorption cuvettes, standard cells, Macro
Suprasil® 300 quartz, spectral range 200-3500 nm, pathlength 1 mm, chamber volume 700 μL
About This Item
Nomenclature NACRES :
SB.32
Matériaux
Suprasil® 300 quartz
Description
2 polished windows
Fabricant/nom de marque
Hellma 404.000-QX
Paramètres
200-3500 nm spectral range
H × l × P
47.5 mm × 23.6 mm × 7.5 m
Volume de la chambre
700 μL
Longueur du trajet optique
1 mm
Description générale
with 2 PTFE stoppers
Conditionnement
Sold as packages containing a single cell (1EA = 1 cell)
Informations légales
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.