Principaux documents
20716
Seringue Hamilton®, GASTIGHT® série 1700, embout Luer
1710LT, volume 100 μL, without needle, Luer Tip, needle size (not included)
1 EA
65,90 €
65,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EA
65,90 €
About This Item
Code UNSPSC :
41122001
eCl@ss :
32070500
65,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Matériaux
glass body
Taille de l'aiguille
(not included)
Caractéristiques
without needle Luer Tip
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Hamilton 81001
L de l'aiguille
, n/a
Volume
100 μL
Description générale
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Application
It was used in calibrating the instrument used for the analysis of dissolved gases by headspace sampling GC with thermal conductivity, flame ionization and FPD detectors.[3]
Informations légales
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Analysis of dissolved gases by headspace sampling gas chromatography with column and detector switching. Preliminary results.
Sarradin, Pierre-Marie, and Jean-Claude Caprais.
Analytical Communications, 33.10, 371-373 (1996)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)