Principaux documents
RPSTA4VP1
Millipore
Rapid Sterility Testing Validation Protocol
A4 Format, Milliflex® Rapid PQ Protocol
1 EACH
1 450,00 €
1 450,00 €
Date d'expédition estimée le22 avril 2025Détails
Demander des informations complémentaires
Changer de vue
1 EACH
1 450,00 €
About This Item
Code UNSPSC :
41116500
Nomenclature NACRES :
NA.85
1 450,00 €
Date d'expédition estimée le22 avril 2025Détails
Demander des informations complémentaires
Niveau de qualité
Compatibilité
for use with Milliflex® Rapid Systems.
Description générale
Rapid Sterility Testing Validation Protocol is designed as a ready to use step by step guide to perform PQ activities of rapid sterility testing using Milliflex® Rapid system. The PQ protocol will enable the QC/QA Labs to initiate the performance qualification after the IQ andOQ have been completed following the respective separate protocol. The guidelines are GMP compliant and follow EP, USP and JP regulatory guidelines.
Application
Rapid Sterility Testing Validation Protocol is used to validate rapid sterility testing procedures with ease and consistent performance while following regulatory guidelines.
Informations légales
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany