Principaux documents
EZSPUMP01
Millipore
Liquid Pump
for use with EZ-Stream® Vacuum Pump
1 EACH
973,00 €
973,00 €
Date d'expédition estimée le15 avril 2025Détails
Demander des informations complémentaires
Changer de vue
1 EACH
973,00 €
About This Item
Code UNSPSC :
41116500
Nomenclature NACRES :
NB.79
973,00 €
Date d'expédition estimée le15 avril 2025Détails
Demander des informations complémentaires
Caractéristiques
repair kit
Fabricant/nom de marque
EZ-Stream®
Application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
Compatibilité
for use with EZ-Stream® Vacuum Pump
Description générale
Liquid Pump is a spare part for EZ-Stream® Vacuum Pump, it is used to replace internal liquid pump in case of malfunction or filtration issue to keep optimal filtration performance.
The design of the system allows fast and easy replacement of this component.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
The kit includes:
  • 1 Liquid Pump.
Application
Liquid Pump is a spare part used for EZ-Stream® Vacuum Pump. The EZ-Stream® Vacuum Pump is used to provide hassle free liquid discard during filtration process.
Informations légales
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany