Documents
Z682454
Sigma-Aldrich
Ace vapor duct tube, for condenser "V" assembly
joint: ST/NS 29/32, L 45 mm, fits Büchi® models R-210/R-215
Synonyme(s) :
Ace vapor duct tube, for condenser "V" assembly
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/32
Fabricant/nom de marque
Büchi equivalent, 46962
Longueur
45 mm
Description générale
The vapor duct tube accessories are for Büchi condenser "V" assemblies, and will fit either Büchi Rotavapor model R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG