Documents
Z680052
MyBlock mini dry bath block
to hold, 15 X 1.5 mL or 2.0 mL conical bottom tubes
Synonyme(s) :
block dry, dry baths, dry blocks, heat blocks
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NC.21
Caractéristiques
to hold 15 X 1.5 mL or 2.0 mL conical bottom tubes
Fabricant/nom de marque
Benchmark Scientific BSH100-1520
Description générale
To fit MyBlock mini dry bath/block heater.
Informations légales
MyBlock is a trademark of Benchmark Scientific, Inc.