Z677450
Camatril™ Velours nitrile flock lined gloves
size L
Synonyme(s) :
reusable glove
370,00 €
About This Item
370,00 €
Produits recommandés
Matériaux
green gloves
green nitrile
Taille
L
Stérilité
non-sterile
Caractéristiques
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Conditionnement
package of 100 ea
Fabricant/nom de marque
KCL 730 - 09
KCL 730
L × épaisseur
300 mm × 0.4 mm
Description générale
Good flexibility, anatomical design and excellent fit ensure a good sense of touch. The flock lining offers additional wearing comfort.
- Ergonomic fit ensures good sensitivity
- Roughened palm for excellent grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- CE compliant