Principaux documents
Z677434
Camatril Velours nitrile flock lined gloves
size S
Synonyme(s) :
reusable glove
100 EA
370,00 €
370,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EA
370,00 €
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
370,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
green gloves
green nitrile
Taille
S
Stérilité
non-sterile
Caractéristiques
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Conditionnement
package of 100 ea
Fabricant/nom de marque
KCL 730 - 07
KCL 730
L × épaisseur
300 mm × 0.4 mm
Description générale
Camatril Velours green nitrile flock lined gloves are manufactured from high quality nitrile ensuring effective chemical resistance for use in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials.
Good flexibility, anatomical design and excellent fit ensure a good sense of touch. The flock lining offers additional wearing comfort.
  • Ergonomic fit ensures good sensitivity
  • Roughened palm for excellent grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Informations légales
Camatril is a trademark of KCL GmbH