Z671983
Hanna portable waterproof dissolved oxygen meter, model HI9142
battery power 4 AA, 500 hr life
About This Item
Code UNSPSC :
41113319
Produits recommandés
Fabricant/nom de marque
Hanna
Description générale
HI 9142 is a rugged, waterproof meter that solves the common problems of field use, such as cold, rain, snow and dust that can damage a meter, rapidly deteriorating its performance and life.
- Waterproof
- Simple to calibrate
- Built-in thermister for temperature management
- Polarographic probe with 4 m cable