Principaux documents
Z654345
Pouring rings for Duran® laboratory bottles
GL 45 neck thread, colorless
Synonyme(s) :
pouring ring
5 EA
19,00 €
19,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
5 EA
19,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
19,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
colorless
Fabricant/nom de marque
DURAN®
Schott
Paramètres
-196-260 °C temp. stability
Raccords
GL 45 neck thread
Description générale
Thanks to its highly resistant materials, the premium cap is optimally suited to utilize in processes which are both chemically and thermally demanding. Due to its high temperature resistance, it is ideal for demanding processes, such as hot-air / dry-heat sterilisation and depyrogenation at 200 °C. Due to the PTFE-coated silicone cap liner, the premium cap provides a reliable and tight seal. Therefore it is ideal for the long term storage and transport of sensitive products. The matching PFA pouring ring permits drip-free pouring is available separately. Replacement PTFE faced silicone cap liners (platinum-cured silicone) are available
Caractéristiques et avantages
Optional pouring rings prevent drips when pouring from Duran bottles.
Informations légales
DURAN is a registered trademark of DWK Life Sciences
Les clients ont également consulté
Slide 1 of 1