Principaux documents
Z153958
Caps for Duran® laboratory bottles
GL 45 neck thread, red PBT (with PTFE seal)
Synonyme(s) :
caps bottles, glass bottles caps
10 EA
83,70 €
83,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
83,70 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
83,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
red PBT (with PTFE seal)
Fabricant/nom de marque
DURAN®
Schott
Paramètres
-45-180 °C temp. stability
Raccords
GL 45 neck thread
Description générale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
Caractéristiques et avantages
  • High Temperature Screw Cap
  • High Performance
Informations légales
DURAN is a registered trademark of DWK Life Sciences