Principaux documents
Z338389
Filtres pour seringue Whatman® GD/XP
Nylon membrane
Synonyme(s) :
Whatman filter, syringe filter, syringe filters
1 PKG
526,00 €
526,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
526,00 €
About This Item
Code UNSPSC :
41104922
Nomenclature NACRES :
NB.24
526,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
Nylon membrane
polypropylene housing
Stérilité
non-sterile
Caractéristiques
Polypropylene prefilter
Conditionnement
pack of 150 ea
Fabricant/nom de marque
Whatman 6970-2504
Whatman Article No. 28420561 (US reference)
Diamètre
25 mm
Volume
≤25 mL
Dimension de pores
0.45 μm
Raccords
female Luer inlet (lock)
male Luer outlet
Adéquation
Suitable for filtering trace metal analysis samples and for particulate laden sample preparation.
Autres remarques
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Informations légales
Whatman is a registered trademark of Cytiva
Remplacé(e)(s) par