Key Documents
Z304751
Quickfit® air/steam inlet tube
diam. 6.5 mm
Synonyme(s) :
MF5
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Fabricant/nom de marque
Quickfit® MF 5
Disponibilité
not available in USA and Canada
Diamètre
6.5 mm
Description générale
Tube with drawn tip for use with thread size 13 screw cap adapters. Order cone/screw-thread adapters for inlet tube separately (see Z302325).
Caractéristiques et avantages
• Manufactured from borosilicate glass 3.3• With integral jacket through which a temperature controlling medium can be circulated• With PTFE screwthread connectors for simple and safe connection of 9mm bore tubing• PTFE screwthread connectors are available as spare parts
Informations légales
Quickfit is a registered trademark of SciLabware Limited