Documents
Z301159
Quickfit® expansion adapter
top female joint: ST/NS 34/35, bottom male joint: ST/NS 19/26
Synonyme(s) :
XA52
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom19/26
Fabricant/nom de marque
Quickfit XA 52
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited