Documents
Z301078
Quickfit® Reduction Adapters
top female joint: ST/NS 34/35, bottom male joint: ST/NS 50/42
Synonyme(s) :
DA58
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom50/42
Fabricant/nom de marque
Quickfit DA 58
Disponibilité
not available in USA and Canada
Description générale
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited