Documents
SYNA201419
Synthware reducing adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 19/22
Synonyme(s) :
Synthware A201419, glass reducing adapter, connecting adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS19/22
joint: ST/NS top14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A201419
Catégories apparentées
Informations légales
Synthware is a trademark of Kemtech America Inc.