Z247219
Coors™ porcelain crucible cover
cap size B, I.D. 39 mm
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
Produits recommandés
Taille du bouchon
B
Conditionnement
pkg of 12 ea
Fabricant/nom de marque
Coorstek 66123
D.I.
39 mm
Description générale
Coors™ porcelain crucible cover is glazed all over except for the rim.
Caractéristiques et avantages
- Ensures great resistance to thermal-shock
- Durable
- Rigid
- Sustained refractory qualities
Autres remarques
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Informations légales
Coors is a trademark of Coorstek, Inc.