Principaux documents
Z232335
Caps for Duran® laboratory bottles
GL 32 neck thread, blue polypropylene (with lip seal)
Synonyme(s) :
caps bottles, glass bottles caps
10 EA
17,70 €
17,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
17,70 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
17,70 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue polypropylene (with lip seal)
Fabricant/nom de marque
DURAN® 292391907
Schott
Paramètres
-40-140 °C temp. stability
Raccords
GL 32 neck thread
Description générale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Informations légales
DURAN is a registered trademark of DWK Life Sciences