Principaux documents
Z176338
Sigma-Aldrich
Air-driven stirrer
Heavy usage, parameter 50-1200 rpm
Synonyme(s) :
air driven stirrers, air mixer, air stirrer
About This Item
Code UNSPSC :
41103801
Nomenclature NACRES :
NB.72
Le produit Z176338 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MODEL G
Disponibilité
not available in EU
Paramètres
50-1200 rpm
Tige de support, diam. × L
9/16 in. × 10 in.
Description générale
  • Explosion-proof, stall-proof, does not overheat
  • Exhaust muffler
  • Brass needle valve with 1/4 in. NPTM outlet controls air intake
  • Mount on standard clamp holders and vertical rods (order separately)
Includes: a screw collet and 3/8 in. diam. × 12 in. L SS stirrer shaft with propeller. A 3-jaw geared chuck may be ordered separately.