Principaux documents
WHAWB100003
Qiagen® Indicating Dessicant
1g packets, pkg of 1000 ea
Synonyme(s) :
FTA archiving, Indicating Desiccant Pack (1000), Whatman FTA, Z747335
1 PKG
438,00 €
438,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
438,00 €
About This Item
Code UNSPSC :
41105500
Nomenclature NACRES :
NB.22
438,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 1000 ea
Fabricant/nom de marque
Qiagen WB100003
Description générale
Desiccant packets (1g) with indicator to ensure that FTA Cards remain dry during transport or storage. A color change from blue to pink indicate absorption of moisture.
Autres remarques
Field of Use : For internal research use only
Informations légales
Qiagen is a registered trademark of Qiagen Group
Les clients ont également consulté
Articles
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Protocoles
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.