WHA77073700
Whatman® 850-DS 8-channel filter plate
PVDF membrane, pore size 0.2 μm, pack of 50 ea
521,00 €
About This Item
521,00 €
Produits recommandés
Matériaux
PVDF membrane
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Cytiva
Dimension de pores
0.2 μm
Description générale
Application
Caractéristiques et avantages
- Automated processing: up to 8 samples simultaneously
- Readily available: in a wide range of pore sizes and materials