Principaux documents
TEM-LC400CU150
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
size 400 mesh, size 150 nm , hole size, box of 25 ×
Synonyme(s) :
Lacey/ Carbon Film - Cu, 400 mesh 150 micron, 25pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-LC400CU150 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
box of 25 ×
Taille
150 nm , hole size
400 mesh
Description générale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.