Principaux documents
TEM-LC400CU
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
size 400 mesh, size 50 nm , hole size, box of 5 ×
Synonyme(s) :
Lacey/Carbon Film, 400 Mesh, Copper, 5pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-LC400CU n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
box of 5 ×
Taille
400 mesh
50 nm , hole size
Description générale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.