Principaux documents
TEM-LC300CUUL
Sigma-Aldrich
Ultrathin Lacey Carbon Supported Copper Grid
size 300 mesh, box of 25
Synonyme(s) :
LACEY CARBON FILM CU 300# ULTRA THIN 25/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-LC300CUUL n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
box of 25
Taille
300 mesh
Description générale
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Copper Grid, 300 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.