Principaux documents
TEM-LC200-AU-150
Sigma-Aldrich
Lacey Carbon Supported Gold Grid
size 200 mesh, hole size 150 μm , Average, box of 25
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-LC200-AU-150 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
box of 25
Taille de l'orifice
150 μm , Average
Taille
200 mesh
Description générale
Lacey Carbon Supported Grid: Gold Grid, 200 mesh. Average hole size is 150 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.