Principaux documents
TEM-FFT400CU25
Sigma-Aldrich
Formvar Supported Copper Thin Bar Square Mesh
size 400 mesh, box of 25
Synonyme(s) :
FORMVAR SUPPORT FILM THIN BAR SQUARE 400 MESH CU GRIDS
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25
Taille
400 mesh
Description générale
A 10 nm thin film of pure formvar resin supported thin bar square mesh grids. Copper grid, 400 mesh.