Principaux documents
TEM-74357
Sigma-Aldrich
Copper Tomography Grid R14C2
pack of 50 ×
Synonyme(s) :
Copper Tomography Grid, 300 mesh, 50pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
pack of 50 ×
Description générale
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.