Principaux documents
SYNS740100
Synthware sublimation apparatus, complete
100 mL, complete
Synonyme(s) :
Synthware S740100, sublimator
1 EA
464,00 €
464,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
464,00 €
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
464,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S740100
Capacité
100 mL
100 mL
Catégories apparentées
Description générale
Two-piece grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer. Condenser has a large top opening for use with dry ice/acetone or liquid nitrogen. Connection between the flask and condenser is made with a flat O-ring joint permitting parts to be gently separated for maximum product recovery. The capacity listed is that below the bottom of the cold finger. Vacuum control is provided by a 0-8mm high vacuum valve. Complete apparatus consists of the condenser, flask, Viton® O-ring
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company