Documents
SYNF62241L
Synthware pressure equalizing addition funnel
1000 mL, joint: ST/NS 24/40, stopcock: PTFE
Synonyme(s) :
Synthware F62241L, dropping funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F62241L
Capacité
1000 mL
1000 mL
Description générale
With 1:5 PTFE stopcock
Informations légales
Synthware is a trademark of Kemtech America Inc.