Principaux documents
SYNF471125A
Synthware Squibb-style separatory funnel with PTFE stopcock and PTFE stopper
125 mL, top joint: ST/NS 14/20
Synonyme(s) :
Synthware F471125A, glass separatory funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS top14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F471125A
Capacité
125 mL
125 mL
Catégories apparentées
Description générale
Supplied with a Glass or PTFE stopper
Informations légales
Synthware is a trademark of Kemtech America Inc.