Documents
SYNC109130
Synthware Vigreux distillation column
joint: ST/NS 19/22, L 130 mm
Synonyme(s) :
Synthware C109130, distilling column, fractional column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C109130
Longueur
130 mm
Description générale
Column length is measured from the highest to lowest indent in the column.
Informations légales
Synthware is a trademark of Kemtech America Inc.