Principaux documents
CLS709801L
Pyrex® VISTA Erlenmeyer flask, narrow-mouth
capacity 1000 mL
Synonyme(s) :
conical flask, conical flasks, erlenmeyer flasks, flask erlenmeyer, Corning® 70980
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le produit CLS709801L n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
borosilicate glass
narrow-mouth
Stérilité
non-sterile
Caractéristiques
cap: no
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 70980-1L
Capacité
1000 mL
Description générale
These narrow mouth Erlenmeyer flasks have uniform wall thickness which provide the proper balance between mechanical strength and thermal shock resistance. Approximate graduations are in durable blue enamel. An extra large marking space is also provided.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.