Principaux documents
BR88675
BRAND® stopcock T-bore with nozzles
for tube I.D. 8 mm
Synonyme(s) :
PTFE, Stopcocks
1 EA
62,50 €
62,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
62,50 €
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
62,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PTFE
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 88675
D.I.
4 mm
Catégories apparentées
Description générale
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG