Principaux documents
BR87624
BRAND® PP beaker with spout, low form
volume 3000 mL
Synonyme(s) :
Beakers, Plastics
4 EA
86,80 €
86,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
4 EA
86,80 €
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
86,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
Description
in addition to ISO 7056
Caractéristiques
graduations
beaded rim and spout
Fabricant/nom de marque
BRAND 87624
Volume
3000 mL
Description générale
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Autres remarques
Browse all plastic beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG