Principaux documents
BR780741
BRAND® screw cap for micro tubes
blue
Synonyme(s) :
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
1000 EA
123,00 €
123,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1000 EA
123,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
123,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue
polypropylene
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 780741
Catégories apparentées
Description générale
The BRAND® Screw Cap for Micro Tubes is designed specifically for use with micro tubes 780730 - 780734 and 780750 - 780754. Made from high-quality polypropylene (PP), these screw caps feature a silicone seal that ensures a secure closure, preventing leaks and contamination. They are suitable for a wide range of applications, withstanding temperatures from -196 °C to +121 °C. Certified as BIO-CERT® PCR QUALITY, these caps are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, making them ideal for sensitive molecular biology applications.
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG