BR704430
Bottle adapters for BRAND® Dispensers and Burettes
GL 45 thread, for bottle thread S 54, polypropylene
Synonyme(s) :
Accessories, Bottle-top Dispenser, Dispenser, Dispensette®
24,20 €
About This Item
24,20 €
Produits recommandés
Matériaux
polypropylene
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 704430
Raccords
GL 45 thread