Principaux documents
BR4630130
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 0.5-5 mL, without recirculation valve
Synonyme(s) :
bottle top dispensers
1 EA
350,00 €
350,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
350,00 €
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
350,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
incremental 0.1 mL
DE-M marking
without recirculation valve
Fabricant/nom de marque
BRAND 4630130
Gamme de volumes
0.5-5 mL
Raccords
(GL 28/S28)
GL 32
GL 38
S 40
Catégories apparentées
Description générale
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.