BR4600140
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 1-10 mL, without recirculation valve
Synonyme(s) :
bottle top dispensers
350,00 €
About This Item
350,00 €
Produits recommandés
Caractéristiques
incremental 0.2 mL
DE-M marking
without recirculation valve
Fabricant/nom de marque
BRAND 4600140
Gamme de volumes
1-10 mL
Raccords
(GL 28/S28)
GL 32
GL 38
S 40
Catégories apparentées
Description générale
Caractéristiques et avantages
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Les clients ont également consulté
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.