Principaux documents
BR4600340
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 1-10 mL, without recirculation valve
Synonyme(s) :
Digital Bottle-Top Dispenser
1 EA
405,00 €
405,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
405,00 €
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
405,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
incremental 0.05 mL
DE-M marking
without recirculation valve
Fabricant/nom de marque
BRAND 4600340
Gamme de volumes
1-10 mL
Raccords
(GL 28/S28)
GL 32
GL 38
S 40
Catégories apparentées
Description générale
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.