BR43338
BRAND® BLAUBRAND® density bottle, Gay-Lussac pattern, calibrated
capacity 100 mL, borosilicate glass
Synonyme(s) :
BLAUBRAND®, Gay-Lussac
About This Item
Produits recommandés
Matériaux
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
Caractéristiques
with cap
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 43338
Capacité
100 mL
Protocoles
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.