BR36401
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 1 mL, polypropylene stopper
Synonyme(s) :
BLAUBRAND®, Volumetric flasks
44,60 €
About This Item
44,60 €
Produits recommandés
Matériaux
polypropylene stopper
transparent borosilicate glass 3.3 flask
Joint
joint ST/NS neck7/16
Caractéristiques
accuracy class A
Conditionnement
pack of 2 ea
Fabricant/nom de marque
BRAND 36401
Capacité
1 mL
Description générale
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.