Principaux documents
BR16520
BRAND® burette clamp
Zinc alloy, nickel-plated for 2 burettes, clamp PVC-coated
Synonyme(s) :
Accessories, Burette clamp, Burettes
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.01
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 16520
Description générale
Burettes up to 50 ml can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible. Very sturdy design, easy to operate. Integral support connector.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG