Principaux documents
BR114720
BRAND® grip stopper for sample and sedimentation tubes
for sample tubes BR114715
Synonyme(s) :
Accessories, Stoppers, Test tubes, Tubes
1000 EA
28,80 €
28,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1000 EA
28,80 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
28,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
low-density polyethylene
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
BRAND 114720
Catégories apparentées
Description générale
The BRAND® Grip Stopper is specifically designed for use with sample and sedimentation tubes 114715, providing a secure and effective sealing solution. Made from neutral PE-LD (low-density polyethylene), this stopper ensures compatibility with various substances while maintaining a tight seal to prevent contamination. Its ergonomic design allows for easy handling and application, making it ideal for laboratory environments. This grip stopper enhances the functionality of your tubes, ensuring safe storage and transport of samples and liquids while facilitating reliable sedimentation processes.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG