Documents
BMSBSWBCT
Benchmark Dry Bath Blocks
to hold, blood collection tubes, 12 x 13 mm and 8 x 16mm
Synonyme(s) :
dry bath block, block heater block, drilled block
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NC.21
Matériaux
aluminum
Caractéristiques
to hold blood collection tubes, 12 x 13 mm and 8 x 16mm
Fabricant/nom de marque
Benchmark Scientific BSWBCT