Principaux documents
BAH369300050
Sterileware sampling spatula
polystyrene, extra long, sterile
50 EA
104,00 €
104,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
50 EA
104,00 €
About This Item
Code UNSPSC :
41122403
Nomenclature NACRES :
NB.35
104,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polystyrene
Stérilité
sterile
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Bel-Art H36930-0050
L totale
356 mm (14 in.)
Longueur du manche
102 mm (4 in.)
Description générale
One-piece spatula is molded from rigid, white polystyrene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.
  • V-shaped spatula is perfect for sampling granulated or semi solid material
  • Sterile and individually packaged
  • One time use averts cross contamination
  • Sharp point will pierce most packaging
Informations légales
Sterileware is a trademark of SP Scienceware, a division of SP