Principaux documents
BAF369100060
SP Bel-Art® Sterileware Pharma Scoop
red polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 100 ea (individually wrapped)
Synonyme(s) :
Lab Scoop, spoon
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
red polystyrene
Stérilité
sterile
Conditionnement
pack of 100 ea (individually wrapped)
Fabricant/nom de marque
Bel-Art F36910-0060
bowl H × L × W
1.125 in. (28 mm) × 2.875 in. (72 mm) × 2 in. (49 mm)
Capacité nominale
60 mL (2 oz)
Description générale
Ideal for sampling for both production work and laboratory use, Pharma Scoops are used by pharmaceutical, food, cosmetic, and chemical companies.
Caractéristiques et avantages
  • High handle location keeps hand further from sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable. These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP